Learn More
Medchemexpress LLC 2-Hydroxycinnamic acid | 614-60-8 | MFCD00004379 | 98.71% | 164.16 | 100 MG
SDP
Catalog No. 5000453024
Encompass_Preferred
2-Hydroxycinnamic acid (Standard) serves as the analytical standard for 2-Hydroxycinnamic acid, intended for research and analytical applications. It is a phenolic acid possessing antimicrobial and antioxidant properties. This analytical standard grade compound is commonly employed in qualitative, quantitative, and methodological research experiments, including HPLC, GC, and MS.
  • Demonstrates antimicrobial activity against *Staphylococcus aureus* without susceptibility to drug resistance.
  • Exhibits inhibitory effects on HIV/SARS-CoV S pseudovirus infection, with an IC50 of 0.3 mM.
  • Possesses neuroprotective activity.
  • Possesses antitumor activity.
Catalog No. 50-004-53024 Supplier Medchemexpress LLC Supplier No. HYW012531R100MG
2-Hydroxycinnamic acid (Standard) serves as the analytical standard for 2-Hydroxycinnamic acid, intended for research and analytical applications. It is a phenolic acid possessing antimicrobial and antioxidant properties. This analytical standard grade compound is commonly employed in qualitative, quantitative, and methodological research experiments, including HPLC, GC, and MS.
  • Demonstrates antimicrobial activity against *Staphylococcus aureus* without susceptibility to drug resistance.
  • Exhibits inhibitory effects on HIV/SARS-CoV S pseudovirus infection, with an IC50 of 0.3 mM.
  • Possesses neuroprotective activity.
  • Possesses antitumor activity.