Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Hydroxydiphenylmethane, 98%, Thermo Scientific™

Catalog No. AC162000250
Change view
Click to view available options
Quantity:
25 g
Packaging:
Plastic bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC162000250 25 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC162000250 Supplier Thermo Scientific Chemicals Supplier No. 162000250

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 28994-41-4
Molecular Formula C13H12O
Molecular Weight (g/mol) 184.238
MDL Number MFCD00002242
InChI Key CDMGNVWZXRKJNS-UHFFFAOYSA-N
Synonym 2-hydroxydiphenylmethane, o-benzylphenol, phenol, 2-phenylmethyl, benzylphenol, 2-benzyl-phenol, 2-phenylmethyl phenol, alpha-phenyl-o-cresol, o-cresol, .alpha.-phenyl, unii-sf555bl24o, 2-benzyl phenol
PubChem CID 24216
IUPAC Name 2-benzylphenol
SMILES C1=CC=C(C=C1)CC2=CC=CC=C2O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 51°C to 55°C
Color Colorless to Pink-Red
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Plastic bottle
Linear Formula C6H5CH2C6H4OH
Quantity 25 g
Formula Weight 184.24
Percent Purity ≥97.5%
Physical Form Crystals
Chemical Name or Material 2-Hydroxydiphenylmethane, 98%
Show More Show Less
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 249-361-8
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.