Learn More
Cayman Chemical 2-Hydroxyestradiol, Purity: ≥95%, CAS: 362-05-0, 10 mg
SDP
Catalog No. 502953196
Encompass_Preferred
A CYP450 metabolite of estradiol with low affinity for estrogen receptors; rapidly metabolized to 2-ME2 with a half-life of approximately ten minutes in rat
Catalog No. 50-295-3196 Supplier Cayman Chemical Supplier No. 1301910 MG
A CYP450 metabolite of estradiol with low affinity for estrogen receptors; rapidly metabolized to 2-ME2 with a half-life of approximately ten minutes in rat