Learn More
Medchemexpress LLC 2-Hydroxyestrone-d4 | 81586-97-2 | 97.9% | 290.39 | 1 MG
SDP
Catalog No. 5000436934
Encompass_Preferred
2-Hydroxyestrone-d4 is the deuterium-labeled form of 2-Hydroxyestrone. 2-Hydroxyestrone (Catecholestrone) acts as a specific receptor-mediated antiestrogenic agent and is recognized for its anticarcinogenic properties.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process.
  • Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-36934 Supplier Medchemexpress LLC Supplier No. HY113251S1MG
2-Hydroxyestrone-d4 is the deuterium-labeled form of 2-Hydroxyestrone. 2-Hydroxyestrone (Catecholestrone) acts as a specific receptor-mediated antiestrogenic agent and is recognized for its anticarcinogenic properties.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process.
  • Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.