Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Hydroxyethyl 2-bromoisobutyrate

Catalog No. 7231505G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
72-315-05G 5 g
72-315-01G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 72-315-05G Supplier Sigma Aldrich Supplier No. 7231505G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Atom Transfer Radical Polymerization (ATRP) initiator for the creation of hydroxy functionalized telechelic polymers. Can be used to modify carboxylate- or isocyanate- modified surfaces, particles or biomolecules.

Specifications

CAS 189324-13-8
Density 1.456 g/mL (at 25°C)
Molecular Formula C6H11BrO3
Linear Formula C6H11BrO3
Refractive Index n20/D 1.476
MDL Number MFCD16621490
Quantity 5 g
Synonym ATRP hydroxy initiator; HEBIB; HO-BriBu
Molecular Weight (g/mol) 211.05
Percent Purity 95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.