Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-hydroxyethyl 4-methylbenzenesulfonate | 42772-85-0 | 216.26 | C9H12O4S | 500 MG
SDP

Catalog No. 5000208541
Encompass_Preferred

Theophylline impurity 9 (2-hydroxyethyl 4-methylbenzenesulfonate) is an analytical reference standard used for identification and quantification of a theophylline-related impurity in research and quality-control workflows.

  • Analytical reference standard for theophylline impurity identification.
  • Cas 42772-85-0.
  • Molecular formula C9H12O4S; molecular weight 216.26 g/mol.
  • Available pack sizes include 100 mg, 250 mg, 500 mg, and 1 g; larger sizes by inquiry.
  • Storage: pure form -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).
  • Batch-specific purity and analytical data provided in the certificate of analysis.

Catalog No. 50-002-08541 Supplier Medchemexpress LLC Supplier No. HYW087424500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Theophylline impurity 9 (2-hydroxyethyl 4-methylbenzenesulfonate) is an analytical reference standard used for identification and quantification of a theophylline-related impurity in research and quality-control workflows.

  • Analytical reference standard for theophylline impurity identification.
  • Cas 42772-85-0.
  • Molecular formula C9H12O4S; molecular weight 216.26 g/mol.
  • Available pack sizes include 100 mg, 250 mg, 500 mg, and 1 g; larger sizes by inquiry.
  • Storage: pure form -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).
  • Batch-specific purity and analytical data provided in the certificate of analysis.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.