Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Hydroxyethyl Acetate, Spectrum™ Chemical
SDP

Catalog No. 18620818 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-620-818 25 mL
18-620-819 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-620-818 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. H146625ML
Add to Cart
Add to Cart

Synonym: Ethylene Glycol Monoacetate, Glycol Monoacetate, 2-Acetoxyethanol

Chemical Identifiers

CAS 542-59-6
Molecular Weight (g/mol) 102.13

Specifications

Quantity 25 mL
Packaging Amber Glass
Boiling Point 128°C
Percent Purity 98%
Grade Ungraded
Chemical Name or Material Ethyl Glycidyl Ether
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.