Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Hydroxyethyl Acetate 60.0+%, TCI America™
SDP

Catalog No. A105925ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A105925ML 25 mL
A1059500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A105925ML Supplier TCI America Supplier No. A105925ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 542-59-6
Molecular Formula C4H8O3
Molecular Weight (g/mol) 104.105
MDL Number MFCD00002864
InChI Key HXDLWJWIAHWIKI-UHFFFAOYSA-N
Synonym Ethylene Glycol Monoacetate, Acetic Acid 2-Hydroxyethyl Ester
PubChem CID 10960
IUPAC Name 2-hydroxyethyl acetate
SMILES CC(=O)OCCO

Specifications

Color Colorless
Boiling Point 182°C
Quantity 25 mL
Formula Weight 104.11
Percent Purity ≥60.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Hydroxyethyl Acetate
EINECSNumber (2)-0738&(2)-0665
RTECSNumber KW7175000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.