Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Hydroxyethyl acrylate

Catalog No. 292818250ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
250 mL
1 L
18 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
29-281-8250ML 250 mL
29-281-818L 18 L
29-281-81L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 29-281-8250ML Supplier Sigma Aldrich Supplier No. 292818250ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

2-Hydroxyethyl acrylate is a hydroxyl group containing vinyl monomer.Porous hydrogels can be prepared by copolymerization of 2-hydroxyethyl acrylate and a cross linking agent. 2-hydroxyethyl acrylate may be used in the synthesis of amphilic block copolymers by nitroxide mediated living radical polymeration. Also, it may be used to prepare tuned poly(hydroxyethyl acrylate) by atom transfer radical polymerization.

Specifications

CAS 818-61-1
Density 1.011 g/mL (at 25°C)
Boiling Point 90°C to 92°C (2 mmHg)
Molecular Formula C5H8O3
Refractive Index n20/D 1.45 (literature)
Linear Formula CH2=CHCOOCH2CH2OH
Quantity 250 mL
MDL Number MFCD00002865
Synonym Ethylene glycol monoacrylate
Molecular Weight (g/mol) 116.12
Percent Purity 96%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.