Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Hydroxyethyl disulfide

Catalog No. 38047450ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
50 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
38-047-450ML 50 mL
38-047-4250ML 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 38-047-450ML Supplier Sigma Aldrich Supplier No. 38047450ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

2-Hydroxyethyl disulfide

Specifications

CAS 1892-29-1
Melting Point 25°C to 27°C
Density 1.261 g/mL (at 25°C)
Boiling Point 158°C to 163°C (3.5 mmHg)
Molecular Formula C4H10O2S2
Linear Formula HOCH2CH2SSCH2CH2OH
Refractive Index n20/D 1.57 (literature)
MDL Number MFCD00002906
Quantity 50 mL
Synonym 2,2 -Dithiodiethanol; Bis(2-hydroxyethyl) disulfide
Molecular Weight (g/mol) 154.25
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.