Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Hydroxyethyl methacrylate

Catalog No. 47702825ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
47-702-825ML 25 mL
47-702-8100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 47-702-825ML Supplier Sigma Aldrich Supplier No. 47702825ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

2-Hydroxyethyl methacrylate (HEMA) is biocompatible in nature. Polymeric hydrogel scaffold can be produced by polymerizing HEMA in water.A polymeric hydrogel for drug delivery. Patent has been applied for the development of artificial cornea or KPro, it is composed of pHEMA (polymerized from HEMA) and poly (methyl methacrylate)(PMMA). HEMA may be used as a macromonomer for the synthesis of 2-hydroxyethylmethacrylate-poly(α-caprolactone) (HEMA-PCL) by coordinated anionic ring opening polymerization (ROP)

Specifications

CAS 868-77-9
Density 1.073 g/mL (at 25°C)
Boiling Point 67°C (3.5 mmHg)
Molecular Formula C6H10O3
Refractive Index n20/D 1.453 (literature)
Linear Formula CH2=C(CH3)COOCH2CH2OH
Quantity 25 mL
MDL Number MFCD00002863
Synonym 1,2-Ethanediol mono(2-methylpropenoate); Glycol methacrylate; HEMA
Molecular Weight (g/mol) 130.14
Percent Purity ≥99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.