Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Grainger TCI 2-Hydroxyethyl Methacrylate, Reagent, 95.0% (GC), 500 g

Catalog No. 502781367
Encompass_Preferred

2-Hydroxyethyl Methacrylate is a reagent-grade laboratory chemical supplied in a 500 g bottle. Stabilized with MEHQ, this compound appears as a colorless to almost colorless clear liquid and is completely miscible in water. It is suitable for diverse laboratory applications requiring precise chemical synthesis or reagent-based reactions.

  • Formula: C6H10O3
  • Molecular weight: 130.14 g/mol
  • Purity: ≥95.0% (GC)
  • Refractive index: 1.45
  • Boiling point: 85°C/5 mmHg
  • Density: 1.07 (20/20°C)
  • Storage: Cool and shaded area (recommended <15°C); light-sensitive
  • Hazard information: Causes skin irritation (H315), serious eye irritation (H319), and may cause an allergic skin reaction (H317)

Catalog No. 50-278-1367 Supplier Grainger Supplier No. 19KT06
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Hydroxyethyl Methacrylate is a reagent-grade laboratory chemical supplied in a 500 g bottle. Stabilized with MEHQ, this compound appears as a colorless to almost colorless clear liquid and is completely miscible in water. It is suitable for diverse laboratory applications requiring precise chemical synthesis or reagent-based reactions.

  • Formula: C6H10O3
  • Molecular weight: 130.14 g/mol
  • Purity: ≥95.0% (GC)
  • Refractive index: 1.45
  • Boiling point: 85°C/5 mmHg
  • Density: 1.07 (20/20°C)
  • Storage: Cool and shaded area (recommended <15°C); light-sensitive
  • Hazard information: Causes skin irritation (H315), serious eye irritation (H319), and may cause an allergic skin reaction (H317)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.