Learn More
Medchemexpress LLC 2-Hydroxyethyl methacrylate | 868-77-9 | 99.1% | 130.14 | 1 KG
SDP
Catalog No. 5000454774
Encompass_Preferred
2-Hydroxyethyl methacrylate (HEMA) is a monomer utilized in the synthesis of various polymers. The polymer PHEMA, derived from 2-Hydroxyethyl methacrylate, finds extensive application in the creation of dental composite materials. It is for research use only.
  • Monomer used in the synthesis of various polymers
  • PHEMA polymer is widely used in dental composite materials
  • Intended for research use only
Catalog No. 50-004-54774 Supplier Medchemexpress LLC Supplier No. HYW0127231KG
2-Hydroxyethyl methacrylate (HEMA) is a monomer utilized in the synthesis of various polymers. The polymer PHEMA, derived from 2-Hydroxyethyl methacrylate, finds extensive application in the creation of dental composite materials. It is for research use only.
  • Monomer used in the synthesis of various polymers
  • PHEMA polymer is widely used in dental composite materials
  • Intended for research use only