Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Hydroxyethyl salicylate, 98+%, Thermo Scientific Chemicals

Catalog No. AC174102500
Change view
Click to view available options
Quantity:
250 mL
1 L
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC174102500 250 mL Glass bottle
AC174100010 1 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC174102500 Supplier Thermo Scientific Chemicals Supplier No. 174102500
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87-28-5
Molecular Formula C9H10O4
Molecular Weight (g/mol) 182.18
MDL Number MFCD00002862
InChI Key LVYLCBNXHHHPSB-UHFFFAOYSA-N
Synonym 2-hydroxyethyl salicylate, glycol salicylate, ethylene glycol monosalicylate, espirosal, glycol monosalicylate, rheumacyl, sarocol, spirosal, glysal, phlogont
PubChem CID 6880
ChEBI CHEBI:86541
IUPAC Name 2-hydroxyethyl 2-hydroxybenzoate
SMILES OCCOC(=O)C1=CC=CC=C1O

Specifications

Melting Point 25°C
Color Colorless
Density 1.2500g/mL
Boiling Point 172°C (12.0 mmHg)
Flash Point 174°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Linear Formula 2-(HO)C6H4CO2CH2CH2OH
Quantity 250 mL
Beilstein 10, 81
Specific Gravity 1.25
Merck Index 15, 4535
Solubility Information Solubility in water: 13g/L (20°C). Other solubilities: very soluble in alcohol,benzene,chloroform,,ether
Formula Weight 182.18
Percent Purity 98+%
Physical Form Oily Liquid or Low Melting Solid
Chemical Name or Material 2-Hydroxyethyl salicylate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 201-737-2

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.