Learn More
Medchemexpress LLC 2-hydroxyfluorene | 2443-58-5 | MFCD00010778 | 99.8% | 182.22 g·mol⁻¹ | C13H10O | 250 MG
SDP
Catalog No. 5000302348
Encompass_Preferred
2-Hydroxyfluorene is a polycyclic aromatic hydrocarbon metabolite used as a research reagent and analytical standard in studies of environmental exposure and metabolism. Molecular formula C13H10O; molecular weight 182.22 g·mol⁻¹. The substance is supplied as a purified solid with storage recommendations to maintain stability.
  • Used as a metabolite and exposure marker in research
  • Suitable as an analytical standard for chromatography and mass spectrometry
  • High purity (99.8%)
  • Provided in small solid quantities for laboratory use
  • Stable when stored under recommended conditions: room temperature under nitrogen; in solution, -80 °C for long-term storage
Catalog No. 50-003-02348 Supplier Medchemexpress LLC Supplier No. 250MGHYW016389
2-Hydroxyfluorene is a polycyclic aromatic hydrocarbon metabolite used as a research reagent and analytical standard in studies of environmental exposure and metabolism. Molecular formula C13H10O; molecular weight 182.22 g·mol⁻¹. The substance is supplied as a purified solid with storage recommendations to maintain stability.
  • Used as a metabolite and exposure marker in research
  • Suitable as an analytical standard for chromatography and mass spectrometry
  • High purity (99.8%)
  • Provided in small solid quantities for laboratory use
  • Stable when stored under recommended conditions: room temperature under nitrogen; in solution, -80 °C for long-term storage