Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-Hydroxyhexanoic acid | 6064-63-7 | 99.9% | C6H12O3 | 1 ML
SDP

Catalog No. 5000433684
Encompass_Preferred

2-Hydroxyhexanoic acid is a metabolite that is elevated in metastatic pancreatic neuroendocrine tumors. It does not promote the proliferation, migration, or invasion of pancreatic neuroendocrine tumor cells and is suitable for research into metastatic pancreatic neuroendocrine tumors.

  • For laboratory chemical use
  • Solid appearance, white to off-white
  • Melting point of 60-62 °C
  • Boiling point of 277.83 °C at 760 mmHg
  • Soluble in water and DMSO (≥ 100 mg/mL)
  • Powder storage at -20 °C for 3 years or 4 °C for 2 years
  • Solution storage at -80 °C for 2 years or -20 °C for 1 year
  • For research use only

Catalog No. 50-004-33684 Supplier Medchemexpress LLC Supplier No. HY7595410MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Hydroxyhexanoic acid is a metabolite that is elevated in metastatic pancreatic neuroendocrine tumors. It does not promote the proliferation, migration, or invasion of pancreatic neuroendocrine tumor cells and is suitable for research into metastatic pancreatic neuroendocrine tumors.

  • For laboratory chemical use
  • Solid appearance, white to off-white
  • Melting point of 60-62 °C
  • Boiling point of 277.83 °C at 760 mmHg
  • Soluble in water and DMSO (≥ 100 mg/mL)
  • Powder storage at -20 °C for 3 years or 4 °C for 2 years
  • Solution storage at -80 °C for 2 years or -20 °C for 1 year
  • For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.