Learn More
Cayman Chemical 2-Hydroxyisocaproic Acid, Purity: ≥98%, CAS: 498-36-2, 1 g
SDP
Catalog No. 502912558
Encompass_Preferred
A metabolite of L-leucine catabolism; is active against a variety of bacteria (IC50s = 0.6-4.5 mg/ml) and several Candida species (IC50s = 18-36 mg/ml); prevents reductions in protein synthesis during the immobilization period and increases protein synthesis and muscle mass during the recovery period in casted animals in a rat model of immobilization-induced muscle atrophy at 50.4 g/kg in the diet
Catalog No. 50-291-2558 Supplier Cayman Chemical Supplier No. 364391 G
A metabolite of L-leucine catabolism; is active against a variety of bacteria (IC50s = 0.6-4.5 mg/ml) and several Candida species (IC50s = 18-36 mg/ml); prevents reductions in protein synthesis during the immobilization period and increases protein synthesis and muscle mass during the recovery period in casted animals in a rat model of immobilization-induced muscle atrophy at 50.4 g/kg in the diet