Learn More
Medchemexpress LLC 2-Hydroxyisoquinolin 250mg
SDP
Catalog No. 5000204660
Encompass_Preferred
Hydroxyisoquinoline-1 3(2H 4H)-dione (compound 6a) is a HIV-1 integrase (IN) and HIV-1 reverse transcriptase (RT) ribonuclease H (RNase H) inhibitor with an IC50 value of 6 32 5 9 M respectively[1]
Catalog No. 50-002-04660 Supplier Medchemexpress LLC Supplier No. 250MGHYW103287
Hydroxyisoquinoline-1 3(2H 4H)-dione (compound 6a) is a HIV-1 integrase (IN) and HIV-1 reverse transcriptase (RT) ribonuclease H (RNase H) inhibitor with an IC50 value of 6 32 5 9 M respectively[1]