Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-Hydroxyl emodin-1-methyl ether | 346434-45-5 | 98.9% | 300.26 | 1 MG
SDP

Catalog No. 5000426555
Encompass_Preferred

2-Hydroxyl emodin-1-methyl ether is an anthraquinone that can be isolated from the seeds of *Cassia obtusifolia*. This compound is for research use only.

  • Appearance: Solid
  • Color: Light yellow to yellow
  • Formula: C16H12O6
  • Structure classification: Quinones, Anthraquinones
  • Initial source: Plants, Caesalpiniaceae, *Cassia obtusifolia* (L.) H. S. Irwin & Barneby

Catalog No. 50-004-26555 Supplier Medchemexpress LLC Supplier No. HYN99611MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Hydroxyl emodin-1-methyl ether is an anthraquinone that can be isolated from the seeds of *Cassia obtusifolia*. This compound is for research use only.

  • Appearance: Solid
  • Color: Light yellow to yellow
  • Formula: C16H12O6
  • Structure classification: Quinones, Anthraquinones
  • Initial source: Plants, Caesalpiniaceae, *Cassia obtusifolia* (L.) H. S. Irwin & Barneby

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.