Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Hydroxymethyl-12-Crown-4

Catalog No. 3826551G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
38-265-51G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. 38-265-51G Supplier Sigma Aldrich Supplier No. 3826551G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

2-Hydroxymethyl-12-crown-4 can be used:To prepare an ester of lasalocid with crown ether to study its complex formation with monovalent cations.To prepare hydrogel adsorbents for potential usage in the recovery of lithium-ions from seawater.As a starting material in the synthesis of metal-free phthalocyanines and metallophthalocyanines.

Specifications

CAS 16419-60-6
Melting Point 162°C to 164°C
Molecular Formula C7H9BO2
Linear Formula CH3C6H4B(OH)2
Refractive Index n20/D 1.480
MDL Number MFCD00093526
Quantity 5 g
Synonym (2-Tolyl)boronic acid; 2-Methyl-1-phenylboronic acid; 2-Methylbenzeneboronic acid; 2-Methylphenylboronic acid; 2-Tolueneboronic acid; o-Boronotoluene; o-Methylphenylboronic acid; o-Tolueneboronic acid
Molecular Weight (g/mol) 206.24 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.