Learn More
Sigma Aldrich 2-(Hydroxymethyl)-5-(isopentyloxy)-4-pyridinol
Shop All MilliporeSigma Sigma Organic Chemistry Products
Description
- 1-methoxypropane can be used as:
- A reactant to synthesize organic nitrates via atmospheric oxidation
- A standard for the calibration curve to quantify thiobarbituric acid reactive substances in proteins
Specifications
Specifications
| Molecular Formula | C11H17NO3 |
| Linear Formula | C11H17NO3 |
| MDL Number | MFCD14552276 |
| Quantity | 5 g |
| Molecular Weight (g/mol) | 211.26 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.