Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-(Hydroxymethyl)anthraquinone | 17241-59-7 | 10 MM 1 ML
SDP

Catalog No. 5000419648
Encompass_Preferred

2-(Hydroxymethyl)anthraquinone is utilized as a photoremovable protecting group (PRPG) to chemically cage sex pheromone, such as (Z)-11-hexadecen-1-ol (the sex pheromone of *Chilo infuscatellussnellen*).

  • Used as a photoremovable protecting group
  • Chemically cages sex pheromones
  • Purity: 97.22%
  • Molecular weight: 238.24
  • Chemical formula: C15H10O3
  • Appearance: Solid
  • Color: Light yellow to yellow
  • Classified as a quinone and anthraquinone
  • Derived from plants, specifically Schisandra chinensis

Catalog No. 50-004-19648 Supplier Medchemexpress LLC Supplier No. HYN750210MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-(Hydroxymethyl)anthraquinone is utilized as a photoremovable protecting group (PRPG) to chemically cage sex pheromone, such as (Z)-11-hexadecen-1-ol (the sex pheromone of *Chilo infuscatellussnellen*).

  • Used as a photoremovable protecting group
  • Chemically cages sex pheromones
  • Purity: 97.22%
  • Molecular weight: 238.24
  • Chemical formula: C15H10O3
  • Appearance: Solid
  • Color: Light yellow to yellow
  • Classified as a quinone and anthraquinone
  • Derived from plants, specifically Schisandra chinensis

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.