Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-(Hydroxymethyl)anthraquinone | 17241-59-7 | 100 MG
SDP

Catalog No. 5000417187
Encompass_Preferred

2-(Hydroxymethyl)anthraquinone is utilized as a photoremovable protecting group (PRPG) to chemically cage sex pheromones, such as (Z)-11-hexadecen-1-ol, the sex pheromone of Chilo infuscatellussnellen.

  • Purity of 97.22%
  • Solid appearance, light yellow to yellow in color
  • Soluble in DMSO at 100 mg/mL (419.74 mM)
  • Recommended storage for powder is -20°C for up to 3 years or 4°C for 2 years
  • Recommended storage in solvent is -80°C for up to 6 months or -20°C for 1 month

Catalog No. 50-004-17187 Supplier Medchemexpress LLC Supplier No. HYN7502100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-(Hydroxymethyl)anthraquinone is utilized as a photoremovable protecting group (PRPG) to chemically cage sex pheromones, such as (Z)-11-hexadecen-1-ol, the sex pheromone of Chilo infuscatellussnellen.

  • Purity of 97.22%
  • Solid appearance, light yellow to yellow in color
  • Soluble in DMSO at 100 mg/mL (419.74 mM)
  • Recommended storage for powder is -20°C for up to 3 years or 4°C for 2 years
  • Recommended storage in solvent is -80°C for up to 6 months or -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.