Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(Hydroxymethyl)tetrahydropyran, 94%

Catalog No. aal1741706
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1741706 5 g
AAL1741714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1741706 Supplier Thermo Scientific Chemicals Supplier No. L1741706
Only null left
Add to Cart

CAS: 100-72-1 | C6H12O2 | 116.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-72-1
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD00006624
InChI Key ROTONRWJLXYJBD-UHFFFAOYSA-N
Synonym tetrahydropyran-2-methanol, 2h-pyran-2-methanol, tetrahydro, 2-hydroxymethyltetrahydropyran, tetrahydropyran-2-carbinol, 2-hydroxymethyl tetrahydropyran, pyran-2-methanol, tetrahydro, tetrahydro-2h-pyran-2-yl methanol, 2-tetrahydropyranilcarbinol, tetrahydropyranyl-2-methanol, 2-methyloltetrahydro-1,4-pyran
PubChem CID 7524
IUPAC Name oxan-2-ylmethanol
SMILES C1CCOC(C1)CO

Specifications

Melting Point <-70°C
Density 1.027
Boiling Point 186°C to 189°C
Flash Point 93°C (199°F)
Refractive Index 1.458
Quantity 5 g
Beilstein 102998
Formula Weight 116.16
Percent Purity 94%
Chemical Name or Material 2-(Hydroxymethyl)tetrahydropyran
Hazard Category H227-H315-H319
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362-P370+P378q-P501c
EINECSNumber 202-882-4
RTECSNumber UQ0175000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.