Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Hydroxymethylbenzothiazole, 98%, Thermo Scientific™

Catalog No. AC358500010
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC358500010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC358500010 Supplier Thermo Scientific Chemicals Supplier No. 358500010
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 37859-42-0
Molecular Formula C8H7NOS
Molecular Weight (g/mol) 165.21
MDL Number MFCD00226293
InChI Key PQXMQZYDBQBWNL-UHFFFAOYSA-N
Synonym benzo d thiazol-2-ylmethanol, 2-benzothiazolemethanol, 2-hydroxymethylbenzothiazole, 1,3-benzothiazol-2-yl-methanol, benzothiazol-2-ylmethan-1-ol, zlchem 107, pubchem14535, benzothiazol-2-yl-methanol, chembl71700, 1,3-benzothiazol-2-yl methanol
PubChem CID 268122
IUPAC Name 1,3-benzothiazol-2-ylmethanol
SMILES OCC1=NC2=CC=CC=C2S1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 95.0°C to 96.0°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 165.22
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2-Hydroxymethylbenzothiazole, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.