Learn More
Cayman Chemical 2-Hydroxyanthraquinone, Purity: ≥98%, CAS: 605-32-3, 10 g
SDP
Catalog No. 502950373
Encompass_Preferred
An anthraquinone with antibacterial and estrogenic activities; active against B. subtilis and B. cereus (MICs = 1.9 and 62.5 µg/ml, respectively); induces ERα activation in a yeast two-hybrid assay at 19 µM
Catalog No. 50-295-0373 Supplier Cayman Chemical Supplier No. 3290610 G
An anthraquinone with antibacterial and estrogenic activities; active against B. subtilis and B. cereus (MICs = 1.9 and 62.5 µg/ml, respectively); induces ERα activation in a yeast two-hybrid assay at 19 µM