Learn More
Medchemexpress LLC 2-Hydroxyoctanoic ac | 617-73-2 | 25mg
SDP
Catalog No. 5000302939
Encompass_Preferred
2-Hydroxyoctanoic acid is a medium chain acyl-CoA synthetase inhibitor with a Ki of 500 M[1]
Catalog No. 50-003-02939 Supplier Medchemexpress LLC Supplier No. 25MGHYW015444
2-Hydroxyoctanoic acid is a medium chain acyl-CoA synthetase inhibitor with a Ki of 500 M[1]