Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-hydroxyoctanoic acid | 617-73-2 | MFCD00014410 | 98.0% | 160.21 g·mol⁻¹ | C8H16O3 | 1 ML
SDP

Catalog No. 5000303703
Encompass_Preferred

2-Hydroxyoctanoic acid is a small-molecule research reagent that inhibits medium-chain acyl-CoA synthetase (reported Ki = 500 μM). It is intended for biochemical and cellular studies and is supplied with documentation for research use.

  • Acts as a medium-chain acyl-CoA synthetase inhibitor (Ki = 500 μM).
  • Chemical formula C8H16O3; molecular weight 160.21 g·mol⁻¹.
  • Purity typically 98.0% by supplier documentation.
  • Available as 10 mM solution in DMSO and as solid quantities (25-500 mg).
  • Intended for research use only; supplied with datasheet and SDS.

Catalog No. 50-003-03703 Supplier Medchemexpress LLC Supplier No. HYW01544410MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

2-Hydroxyoctanoic acid is a small-molecule research reagent that inhibits medium-chain acyl-CoA synthetase (reported Ki = 500 μM). It is intended for biochemical and cellular studies and is supplied with documentation for research use.

  • Acts as a medium-chain acyl-CoA synthetase inhibitor (Ki = 500 μM).
  • Chemical formula C8H16O3; molecular weight 160.21 g·mol⁻¹.
  • Purity typically 98.0% by supplier documentation.
  • Available as 10 mM solution in DMSO and as solid quantities (25-500 mg).
  • Intended for research use only; supplied with datasheet and SDS.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.