Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-hydroxyoctanoic acid | 617-73-2 | MFCD00014410 | 98.0% | 160.21 g/mol | C8H16O3 | 100 MG
SDP

Catalog No. 5000302621
Encompass_Preferred

2-hydroxyoctanoic acid is a small-molecule inhibitor of medium-chain acyl-CoA synthetase (reported Ki ≈ 500 μM) used for biochemical and metabolic research. It is supplied as a white to off-white solid with reported purity of about 98% and has molecular formula C8H16O3 (MW 160.21 g/mol). Consult the manufacturer's datasheet for handling, storage, and safety information.

  • Inhibits medium-chain acyl-CoA synthetase (Ki ≈ 500 μM).
  • White to off-white solid suitable for analytical and biological studies.
  • Purity reported at 98.0% for reproducible results.
  • Molecular formula C8H16O3; molecular weight 160.21 g/mol.
  • Soluble in DMSO; manufacturer provides recommended in vivo formulations.
  • Stable as powder when refrigerated; follow datasheet for storage of solutions.

Catalog No. 50-003-02621 Supplier Medchemexpress LLC Supplier No. HYW015444100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

2-hydroxyoctanoic acid is a small-molecule inhibitor of medium-chain acyl-CoA synthetase (reported Ki ≈ 500 μM) used for biochemical and metabolic research. It is supplied as a white to off-white solid with reported purity of about 98% and has molecular formula C8H16O3 (MW 160.21 g/mol). Consult the manufacturer's datasheet for handling, storage, and safety information.

  • Inhibits medium-chain acyl-CoA synthetase (Ki ≈ 500 μM).
  • White to off-white solid suitable for analytical and biological studies.
  • Purity reported at 98.0% for reproducible results.
  • Molecular formula C8H16O3; molecular weight 160.21 g/mol.
  • Soluble in DMSO; manufacturer provides recommended in vivo formulations.
  • Stable as powder when refrigerated; follow datasheet for storage of solutions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.