Learn More
Medchemexpress LLC 2-Hydroxypalmitic acid | 764-67-0 | 1 ML
SDP
Catalog No. 5000418707
Encompass_Preferred
2-Hydroxypalmitic acid is an intermediate in the phytosphingosine metabolic pathway.
  • Exclusively incorporated into sphingolipids
  • Promotes the production of odd-carbon phosphatidylcholine species
  • Applicable to studies of Saccharomyces cerevisiae infection
Catalog No. 50-004-18707 Supplier Medchemexpress LLC Supplier No. HYN781410MM/1ML
2-Hydroxypalmitic acid is an intermediate in the phytosphingosine metabolic pathway.
  • Exclusively incorporated into sphingolipids
  • Promotes the production of odd-carbon phosphatidylcholine species
  • Applicable to studies of Saccharomyces cerevisiae infection