Learn More
Apexbio Technology LLC 2-Hydroxypalmitic acid 764-67-0 100mg
SDP
Catalog No. 5000095919
Encompass_Preferred
2-Hydroxypalmitic acid (CAS 764-67-0) is a hydroxy fatty acid classified as a derivative of long-chain saturated fatty acids Predominantly found as a minor constituent in certain biological contexts it serves as a metabolic intermediate in lipid metabolism The presence of a hydroxyl group at the C-2 position influences its integration into cellular membranes potentially affecting membrane architecture and associated signaling pathways Owing to these properties 2-hydroxypalmitic acid is utilized in research focused on lipidomics membrane biology and studies of lipid-mediated cell signaling processes
Catalog No. 50-000-95919 Supplier Apexbio Technology LLC Supplier No. A9919100
2-Hydroxypalmitic acid (CAS 764-67-0) is a hydroxy fatty acid classified as a derivative of long-chain saturated fatty acids Predominantly found as a minor constituent in certain biological contexts it serves as a metabolic intermediate in lipid metabolism The presence of a hydroxyl group at the C-2 position influences its integration into cellular membranes potentially affecting membrane architecture and associated signaling pathways Owing to these properties 2-hydroxypalmitic acid is utilized in research focused on lipidomics membrane biology and studies of lipid-mediated cell signaling processes