Learn More
Medchemexpress LLC HY-W015302 100mg Medchemexpress, 2-Aminobenzenesulfonic acid CAS:88-21-1 Purity:>98%
SDP
Catalog No. 501970708
Encompass_Preferred
Medchemexpress, HY-W015590 100mg 2-Hydroxyphenylacetic acid CAS:614-75-5 2-Hydroxyphenylacetic acid is a potential biomarker for the food products, and found to be associated with phenylketonuria (PKU). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0708 Supplier Medchemexpress LLC Supplier No. HYW015302100MG
Medchemexpress, HY-W015590 100mg 2-Hydroxyphenylacetic acid CAS:614-75-5 2-Hydroxyphenylacetic acid is a potential biomarker for the food products, and found to be associated with phenylketonuria (PKU). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.