Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Hydroxyphenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. H11845G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H1184-5G 5 g
H11841G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H1184-5G Supplier TCI America Supplier No. H11845G
Add to Cart
Add to Cart

Specifications

CAS 89466-08-0
Melting Point 185°C
Molecular Formula C6H7BO3
Quantity 5 g
MDL Number MFCD01074581
Synonym 2-hydroxyphenyl boronic acid, 2-hydroxybenzeneboronic acid, o-hydroxyphenylboronic acid, 2-boronophenol, 2-hydroxyphenyl boranediol
InChI Key YDMRDHQUQIVWBE-UHFFFAOYSA-N
SMILES B(C1=CC=CC=C1O)(O)O
IUPAC Name (2-hydroxyphenyl)boronic acid
Molecular Weight (g/mol) 137.929
PubChem CID 2773454
Formula Weight 137.93
Physical Form Crystalline Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.