Learn More
Medchemexpress LLC HY-101103 5g , (2-Hydroxypropyl)-β-cyclodextrin CAS:128446-35-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1011035G
Encompass_Preferred
HY-101103 5g (2-Hydroxypropyl)-β-cyclodextrin CAS: 128446-35-5 (2-Hydroxypropyl)-β-cyclodextrin is a widely used drug delivery vehicle to improve the stability and bioavailability. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9285