Learn More
Cayman Chemical 2-Hydroxypropyl-β-cyclodextrin, 128446-35-5, 1 g
SDP
Supplier:  Cayman Chemical 161691
Encompass_Preferred
A cyclic oligosaccharide containing seven D-(+)-glucopyranose units that is widely used to improve the aqueous solubility of various compounds, especially those containing a phenyl group
Catalog No. NC1387094