Learn More
Sigma Aldrich Fine Chemicals Biosciences (2-Hydroxypropyl)-β-cyclodextrin, 128446-35-5, MFCD00069372, 100 G
Supplier:  Sigma Aldrich Fine Chemicals Biosciences H107100G
Encompass
Powder. (2-Hydroxypropyl)-β-cyclodextrin has been used: to investigate its effects on cholesterol homeostasis, for the kindling procedure in mice and hippocampal electrographic analysis, and to improve poor prepulse inhibition (PPI) in mice.
Catalog No. NC1033414