Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (2-Hydroxypropyl)-Beta-Cyclodextrin

Catalog No. 77890710KG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
100 g
10 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
77-890-710KG 100 g
77-890-7500G 10 kg
77-890-7100G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 77-890-710KG Supplier Sigma Aldrich Supplier No. 77890710KG
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

The solubility of lipophilic drugs increases linearly with the concentration of hydroxypropyl-β-cyclodextrin (HBC) in aqueous solution because of the complex between HBC and the drug. This guest-host type complex is formed between the drug and the non-polar cavity in the HBC that results in enhanced solubility. Solutions may be lyophilized to produce freely soluble powders. Non-toxic in rabbits and mice.Cavasol is a registered trademark of Wacker Chemie AG

Specifications

CAS 174899-83-3
Quantity 100 g
Synonym Cavasol(R) W7 HP
Biological Activity Nervous System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.