Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (2-Hydroxypropyl)-α-cyclodextrin | 128446-33-3 | 1180 | 100 MG
SDP

Catalog No. 5000391719
Encompass_Preferred

(2-Hydroxypropyl)-α-cyclodextrin (MW 1180) is a cyclic oligosaccharide derivative that acts as a solubilizer in various experiments. It is intended for research use only.

  • Acts as a solubilizer in various experiments.
  • Cyclic oligosaccharide derivative.
  • Available in various quantities (e.g., 50 mg, 100 mg, 250 mg, 500 mg, 1 g).
  • Solid appearance, white to off-white color.
  • Storage: Powder at -20°C for 3 years.
  • Solubility in H2O: 100 mg/mL (needs ultrasonic).

Catalog No. 50-003-91719 Supplier Medchemexpress LLC Supplier No. HY166519100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(2-Hydroxypropyl)-α-cyclodextrin (MW 1180) is a cyclic oligosaccharide derivative that acts as a solubilizer in various experiments. It is intended for research use only.

  • Acts as a solubilizer in various experiments.
  • Cyclic oligosaccharide derivative.
  • Available in various quantities (e.g., 50 mg, 100 mg, 250 mg, 500 mg, 1 g).
  • Solid appearance, white to off-white color.
  • Storage: Powder at -20°C for 3 years.
  • Solubility in H2O: 100 mg/mL (needs ultrasonic).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.