Learn More
Cayman Chemical 2-Hydroxypropyl--cyclodextrin, 128446-35-5, 5 g
SDP
Supplier:  Cayman Chemical 161695
Encompass_Preferred
Molecular Formula (C6H9O5)7(C3H7O), Purity ≥95%, Formula Weight 1541.6, Synonyms: 2-hydroxypropyl ethers β-cyclodextrin, HP-β-CD; A cyclic oligosaccharide containing seven D-(+)-glucopyranose units that is widely used to improve the aqueous solubility of various compounds, especially those containing a phenyl group.
Catalog No. NC1129927