Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (2-Hydroxypropyl)-Gamma-Cyclodextrin

Catalog No. 779229100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
77-922-9100G 100 g
77-922-9500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 77-922-9100G Supplier Sigma Aldrich Supplier No. 779229100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(2-Hydroxypropyl)-+a-cyclodextrin (HP-+a-CD) is a hydrophilic compound that can be used as a complexing agent to increase the aqueous solubility and dissolution rate of the drug molecule.HP-+a-CD can be utilized to improve the aqueous solubility and dissolution rate of zolpidem (ZP) upon complexation. ZP-HP-+a-CD complexes showed improved ataxic activity. It is also used to enhance the wound healing property of curcumin by increasing the water solubility.Cavasol is a registered trademark of Wacker Chemie AG

Specifications

CAS 17465-86-0
Quantity 100 g
Synonym Cavasol(R) W8 HP; HGC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.