Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (2-Hydroxypropyl)-Gamma-Cyclodextrin

Catalog No. 39070425G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
39-070-425G 25 g
39-070-45G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 39-070-425G Supplier Sigma Aldrich Supplier No. 39070425G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(2-Hydroxypropyl)-+a-cyclodextrin belongs to the class of cyclodextrin derivatives, which exhibit a greater extent of solubility in water compared to their parent compounds. It is widely used in parenteral formulations and solid dosage forms to aid in the improvement of their stability, dissolution, bioavailability, etc.(2-Hydroxypropyl)-+a-cyclodextrin may be used:As a mobile phase additive in the study of the host-guest interaction with organic low molecular mass compounds prior to their quantification using reversed phase-high performance liquid chromatography (RP-HPLC) technique.As a chiral surfactant for the analysis of econazole by micellar electrokinetic chromatography (MEKC).As an analytical standard for the determination of the analyte in biological samples by HPLC.As a chiral selector for the identification of propiconazole by MEKC.

Specifications

CAS 128446-34-4
Quantity 25 g
Synonym HGC
Molecular Weight (g/mol) ∼1,580 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.