Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Hydroxypyridine-4-carboxylic acid, 98%

Catalog No. AAH6424122
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6424122 100 g
AAH6424106 5 g
AAH6424114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6424122 Supplier Thermo Scientific Chemicals Supplier No. H6424122

missing translation for 'validMainframeMsg'

CAS: 22282-72-0 | C6H5NO3 | 139.11 g/mol

Used in the synthesis of selective inhibitors of neuropeptide Y Y5 inhibiting food intake. Also used in the optimization of p-subsite residues of HIV protease inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used in the synthesis of selective inhibitors of neuropeptide Y Y5 inhibiting food intake. Also used in the optimization of p-subsite residues of HIV protease inhibitors.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22282-72-0
Molecular Formula C6H5NO3
Molecular Weight (g/mol) 139.11
MDL Number MFCD01318566
InChI Key BXHCJLRTXPHUGH-UHFFFAOYSA-N
Synonym 2-hydroxyisonicotinic acid, 2-hydroxy-4-pyridinecarboxylic acid, 2-oxo-1,2-dihydropyridine-4-carboxylic acid, 2-hydroxypyridine-4-carboxylic acid, 2-hydroxy-isonicotinic acid, 2-hydroxyisonicotinicacid, 4-pyridinecarboxylic acid, 1,2-dihydro-2-oxo, pubchem16780, acmc-209yjf, 2-hydroxy isonicotinic acid
PubChem CID 280756
SMILES OC(=O)C1=CC(=O)NC=C1

Specifications

Quantity 100 g
Beilstein 115859
IUPAC Name 2-oxo-1H-pyridine-4-carboxylic acid
Formula Weight 139.11
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 2-Hydroxypyridine-4-carboxylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.