Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Hydroxypyrimidine hydrochloride

Catalog No. H5740825G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H5740825G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H5740825G Supplier Sigma Aldrich Supplier No. H5740825G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

2-Hydroxypyrimidine hydrochloride

Specifications

CAS 38353-09-2
Melting Point 200°C to 205°C (dec.) (lit.)
Molecular Formula C4H4N2O -+ HCl
Linear Formula C4H4N2O -+ HCl
MDL Number MFCD00012781
Quantity 1 g
Synonym 2(1 H)-Pyrimidinone hydrochloride; 2-Pyrimidinol hydrochloride
Molecular Weight (g/mol) 132.55
Percent Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.