Learn More
Medchemexpress LLC 2-Hydroxyquinoline | 59-31-4 | 145.16 | 10 G
SDP
Catalog No. 5000447323
Encompass_Preferred
2-Hydroxyquinoline is an inhibitor of α-glucosidase and α-amylase with IC50 values of 64.4 μg/mL and 130.5 μg/mL, respectively. It can be used in the study of diabetes. This product is for research use only and has not been fully validated for medical applications.
  • Inhibitor of α-glucosidase and α-amylase
  • Suitable for diabetes studies
  • High purity product
Catalog No. 50-004-47323 Supplier Medchemexpress LLC Supplier No. HYW00559810G
2-Hydroxyquinoline is an inhibitor of α-glucosidase and α-amylase with IC50 values of 64.4 μg/mL and 130.5 μg/mL, respectively. It can be used in the study of diabetes. This product is for research use only and has not been fully validated for medical applications.
  • Inhibitor of α-glucosidase and α-amylase
  • Suitable for diabetes studies
  • High purity product