Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-Hydroxyquinoline | 59-31-4 | 145.16 | 25 G
SDP

Catalog No. 5000449990
Encompass_Preferred

2-Hydroxyquinoline is an inhibitor of α-glucosidase and α-amylase, with IC50 values of 64.4 μg/mL and 130.5 μg/mL respectively. This off-white to light yellow solid can be utilized in diabetes research.

  • Inhibitor of α-glucosidase (IC50: 64.4 μg/mL)
  • Inhibitor of α-amylase (IC50: 130.5 μg/mL)
  • Off-white to light yellow solid
  • Soluble in DMSO at 100 mg/mL
  • Purity (HPLC): 99.44%
  • Stable under recommended storage conditions

Catalog No. 50-004-49990 Supplier Medchemexpress LLC Supplier No. 25GHYW005598
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Hydroxyquinoline is an inhibitor of α-glucosidase and α-amylase, with IC50 values of 64.4 μg/mL and 130.5 μg/mL respectively. This off-white to light yellow solid can be utilized in diabetes research.

  • Inhibitor of α-glucosidase (IC50: 64.4 μg/mL)
  • Inhibitor of α-amylase (IC50: 130.5 μg/mL)
  • Off-white to light yellow solid
  • Soluble in DMSO at 100 mg/mL
  • Purity (HPLC): 99.44%
  • Stable under recommended storage conditions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.