Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-Hydroxyquinoline | 59-31-4 | 145.16 | 50 G
SDP

Catalog No. 5000449288
Encompass_Preferred

2-Hydroxyquinoline is an inhibitor of α-glucosidase and α-amylase. With IC50 values of 64.4 μg/mL and 130.5 μg/mL respectively, it is utilized in the study of diabetes.

  • Inhibits α-glucosidase (IC50: 64.4 μg/mL)
  • Inhibits α-amylase (IC50: 130.5 μg/mL)
  • Used in diabetes research
  • Off-white to light yellow solid
  • Soluble in DMSO (100 mg/mL)
  • Store powder at -20°C for 3 years or 4°C for 2 years
  • Store in solvent at -80°C for 6 months or -20°C for 1 month

Catalog No. 50-004-49288 Supplier Medchemexpress LLC Supplier No. HYW00559850G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Hydroxyquinoline is an inhibitor of α-glucosidase and α-amylase. With IC50 values of 64.4 μg/mL and 130.5 μg/mL respectively, it is utilized in the study of diabetes.

  • Inhibits α-glucosidase (IC50: 64.4 μg/mL)
  • Inhibits α-amylase (IC50: 130.5 μg/mL)
  • Used in diabetes research
  • Off-white to light yellow solid
  • Soluble in DMSO (100 mg/mL)
  • Store powder at -20°C for 3 years or 4°C for 2 years
  • Store in solvent at -80°C for 6 months or -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.