Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Imidazolidinethione, 98%

Catalog No. AC122030010
Change view
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Glass bottle
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC122030010 1 kg Plastic bottle
AC122032500 250 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC122030010 Supplier Thermo Scientific Chemicals Supplier No. 122030010
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 96-45-7 | C3H6N2S | 102.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 96-45-7
Molecular Formula C3H6N2S
Molecular Weight (g/mol) 102.16
MDL Number MFCD00005276
InChI Key PDQAZBWRQCGBEV-UHFFFAOYSA-N
Synonym 2-imidazolidinethione, ethylenethiourea, ethylene thiourea, 2-mercaptoimidazoline, imidazolidinethione, n,n'-ethylenethiourea, mercazin i, warecure c, rhenogran etu, mercaptoimidazoline
PubChem CID 2723650
ChEBI CHEBI:34750
IUPAC Name imidazolidine-2-thione
SMILES S=C1NCCN1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 197.0°C to 201.0°C
Color White
Flash Point 252°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Plastic bottle
Quantity 1 kg
Beilstein 24, 4
Merck Index 15, 3858
Solubility Information Solubility in water: 19g/L (20°C). Other solubilities: 440g/L water (90°C), moderately soluble in methanol, ethanol, ethylene, glycol and pyridine, insoluble in acetone, ether, chloroform, benzene, and ligroin
Formula Weight 102.15
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2-Imidazolidinethione, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Suspected of causing cancer.
May damage the unborn child.
Causes damage to organs through prolonged or repeated exposure if inhaled.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wash face,hands and any exposed skin thoroughly after handling.
EINECSNumber 202-506-9
RTECSNumber NI9625000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.