Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-imidazolidinone, 1-phenyl-3-(2-piperidinylmethyl)-, hydrochloride | 1257326-24-1 | 98.3% | 295.81 g/mol | C15H22ClN3O | 100 MG
SDP

Catalog No. 5000246913
Encompass_Preferred

Dopamine D2 receptor agonist-3 is a small-molecule research compound described as a selective dopamine D2 receptor partial agonist and a dopamine D3 receptor antagonist. Supplied as the hydrochloride salt in solid form, it is intended for in vitro and preclinical receptor pharmacology studies; not for human use.

  • Selective D2 partial agonist (pEC50 = 8.3)
  • D3 antagonist activity (pEC50 < 5.5)
  • Molecular formula C15H22ClN3O; molecular weight 295.81 g/mol
  • Purity ≥ 98.3% (LCMS)
  • Supplied as hydrochloride salt, solid form
  • Available in milligram pack sizes for research use
  • Intended for in vitro and preclinical research; not for human use

Catalog No. 50-002-46913 Supplier Medchemexpress LLC Supplier No. HY103418100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Dopamine D2 receptor agonist-3 is a small-molecule research compound described as a selective dopamine D2 receptor partial agonist and a dopamine D3 receptor antagonist. Supplied as the hydrochloride salt in solid form, it is intended for in vitro and preclinical receptor pharmacology studies; not for human use.

  • Selective D2 partial agonist (pEC50 = 8.3)
  • D3 antagonist activity (pEC50 < 5.5)
  • Molecular formula C15H22ClN3O; molecular weight 295.81 g/mol
  • Purity ≥ 98.3% (LCMS)
  • Supplied as hydrochloride salt, solid form
  • Available in milligram pack sizes for research use
  • Intended for in vitro and preclinical research; not for human use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.