Learn More
Medchemexpress LLC 2-imidazolidinone, 1-phenyl-3-(2-piperidinylmethyl) hydrochloride | 1257326-24-1 | 98.3% | 295.81 g/mol | C15H22ClN3O | 50 MG

Supplier: Medchemexpress LLC HY10341850MG
Dopamine D2 receptor agonist-3 is a research compound described as a selective dopamine D2 receptor partial agonist and a dopamine D3 receptor antagonist. It is supplied as the hydrochloride salt and is intended for receptor pharmacology and neuroscience research.
- Selective D2 receptor partial agonist and D3 receptor antagonist.
- Hydrochloride salt, white to off-white solid.
- Purity 98.3%.
- Molecular formula C15H22ClN3O; molecular weight 295.81 g/mol.
- Suitable for in vitro and in vivo receptor pharmacology studies.
- Commonly available in small research sizes (5-500 mg).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.