Learn More
Sigma Aldrich 2-Imidazolidone-4-carboxylic acid
Shop All MilliporeSigma Sigma Organic Chemistry Products
Description
- 2-ethylhexyl thioglycolate can be used:
- In the selective extraction of indium(iii) and gallium(iii)
- As a reagent in the synthesis of conjugated polymers based on thienoquinoidal building block
Specifications
Specifications
| CAS | 41371-53-3 |
| Melting Point | 184°C to 186°C (dec.) (lit.) |
| Molecular Formula | C4H6N2O3 |
| Linear Formula | C4H6N2O3 |
| MDL Number | MFCD00005258 |
| Quantity | 500 mL |
| Synonym | 2-Oxo-4-imidazolidinecarboxylic acid |
| Molecular Weight (g/mol) | 130.1 |
| Percent Purity | 95% |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.